Quality Products from Sheldon Manufacturing
Sheldon Manufacturing provides a variety of innovative equipment for life science, pharmaceutical, biomedical, environmental, and industrial applications. Products include incubators, ovens, water and bead baths, and anaerobic chambers. Sheldon Manufacturing markets a complete line of products under the SHEL LAB and BACTRON brands designed to fit your needs.
Microbiological Incubators
Designed for simplicity and durability, SHEL LAB Microbiological Incubators withstand constant use without losing atmospheric control.
B.O.D. Thermoelectric Cooled Incubators
SHEL LAB refrigerated incubators excel at biological oxygen demand (B.O.D) applications.
BACTRON Anaerobic Chambers
Anaerobic chambers are designed to control atmospheric conditions to protect materials sensitive to oxygen or fulfill special contamination needs.
Forced Air High Performance Ovens
SHEL LAB Horizontal airflow ensures uniform heating profiles and powerful heating elements provide fast heat-up and recovery to ensure rapid drying
BACTRON Anaerobic Chambers
BACTRON chambers are total anaerobic process workstations. Samples can be prepared, cultured, and inspected in the oxygen-free chamber and standard incubator. Unlike anaerobic jars and bags, which unavoidably expose samples to oxygen, BACTRON units provide complete anaerobic environments for all your applications.
- Ergonomic arm ports provide a comfortable work space
- Integraded vacuum pump for quieter operation
- 60% faster airlock cycle with 3,4 or 5 auto cycle iterations
- Auto-sleeve cycle with integrated HEPA filter, 40% faster sleeve cycle
- Low gas supply audible and visual alarm
- Single button auto-commissioning
BACTRON Anaerobic Chamber, 300 Plate Capacity
BACTRON300 delivers optimum productivity in a compact space.
BACTRON Anaerobic Chamber, 600 Plate Capacity
BACTRON chambers allow efficient and dexterous, glove-free handling and inspection of samples.
BACTRON Anaerobic Chamber, 900 Plate Capacity
With anaerobic chambers, productivity is further improved by an up to 50% reduction in sample turnaround time.
BactronEZ Anaerobic Chamber, Compact, 300 Plate Capacity
The BactronEZ is an ideal first anaerobic chamber or replacement chamber for users who desire to upgrade their existing chamber.