Inquarts
Small lead pellets with definite amount of silver.
May be added to crucible before fusion, or tapped into cleaned lead buttons before cupellation.
VWR offers a wide assortment of products and brands in this category. If you can not find the product or brand that you are looking for in this listing, please use the general search field in the top-right corner of the screen to search the entire VWR portfolio.