Air-Tite Syringe & Needle Products
Since 1926, Air-Tite Products has specialized in high-grade disposable and reusable hypodermic supplies. Air-Tite supplies VWR with quality products at economical prices for your particular application, whether it is medical, laboratory, industrial, or veterinary use.
As the primary importer for several companies and as a value-added reseller for many others, Air-Tite provides quality products "overlooked" by some larger suppliers, utilizing purchase volume to pass savings onto you. Additionally, Air-Tite Products functions as a project manager for unique and custom solutions.
Air-Tite syringes and needles for the lab
Lab-ready quality meets economy in Air-Tite brand syringes and needles.
HSW® Norm-Ject® syringes
High-quality, 2-part syringes – all plastic, no rubber, no silicone oil. Purity in a disposable syringe!
HSW® Soft-Ject® syringes
German-engineered 3-part syringes – Luer lock and Luer slip from 1mL to 50mL.
Hypodermic and customizable needles
Air-Tite offers one of the widest selections of needles from multiple suppliers.