HALYARD* Cleanroom & Life Science PPE
Cleanroom and Life Science professionals need suppliers they can depend on to deliver high-quality products that meet their specific needs. Owens & Minor, a leader in global healthcare solutions, offers a comprehensive portfolio of Personal Protective Equipment (PPE), including HALYARD* branded PUREZERO* Cleanroom and Life Science Nitrile Gloves and Masks. The HALYARD* product brand produces and supplies PPE that lead with the company’s quality and innovation, which stems from 140-plus years of industry expertise and supply reliability. 
PUREZERO* Cleanroom Gloves
Accelerator-free1 portfolio offered in three non-sterile and two sterile cleanroom gloves to address the needs of pharmaceutical, medical device, microelectronics and semiconductor manufacturing. 
*1 Not fromulated with these commonly used vulcanizing chemicals: Sulfur, Thiurams, Thiaxoles, Guanidines and Carbonates
PUREZERO* Life Science Gloves
Designed for use in laboratories, research, and clean environments with applications in pharmaceutical, med device manufacturing, biotech and food processing/handling.
PUREZERO* Cleanroom Masks
Made of 3 layers, provides high BFE, offered in pouch and pleated syles and features an exclusive SO SOFT* biocomponent technology.
Other PPE and Sterilization Wrap
HALYARD* offers a full line of PPE for controlled environments. HALYARD* Sterilization Wrap is offered in a variety weights and sizes.