Elements featured suppliers and products
As seen in our Elements publication, the products below represent the variety of high-quality, high performing products and service solutions you need in your everyday work.
Elements
The elements publication from Avantor is your one-stop resource for optimizing research workflows. From consumables, instruments, lab equipment & furniture to expert process insights – you will find flexible customized solutions that propel your research from discovery to delivery.
Crystal Industries
AIREA™ CO2 Incubator with 7" color touchscreen and remote WI-FI control
The air jacketed, dry wall, stackable AIREA CO₂ Incubator with advanced controls and smart features is designed to provide the best support for a variety of cell culture applications with precise control of temperature and CO₂ concentration. Smart capabilities in the intuitive and innovative color touchscreen user interface and remote monitoring accessory simplify its use.
View all Crystal Industries products
ELGA
ELGA LabWater Purification systems
PURCHASE AN ELGA LABWATER SYSTEM AND RECEIVE A SECOND PURIFICATION PACK FREE! ELGA specializes in delivering water purification systems for research, science and clinical environments. Our comprehensive range of pure and ultrapure water (Type I, II, III) purification systems offer several features and benefits including a wide performance range from just a few liters to thousands of liters of water per day.
Search for Promo Code 5188 at vwr.com/promotions!
HORIZON
Corepoint Scientific™ Humidity and Temperature Stability Chambers
Ensure the quality and durability of your products with Corepoint® Scientific Humidity Stability Chambers. These chambers provide a controlled humid environment to deliver superior testing backed by FDA and ICH standards. Improve product quality, test packaging, increase shelf life, and enhance safety with these chambers. With natural refrigerants and ultrasonic humidification, these units feature some of the most advanced technology in the industry.
ELMA
Elmasonic Select ultrasonic cleaners
Dependable, state-of-the-art Elma ultrasonic cleaners are made in Germany. Dynamic mode cleans thoroughly at high intensity. Eco mode is quiet, gentle, and energy-saving. Mix, disperse, and dissolve samples in Pulse mode. Save 4 programs for quick call up and reproducible results. Visit our e-commerce platform to view additional sizes.
View all Elma Corporation products
PHCbi
PHCbi Brand VIP® ECO SMART and FrostLess ultra low temperature freezers
ULT technology innovation in action. The VIP® ECO SMART features next-generation engineering to deliver industry-leading energy efficiency and enhanced security. It uses 30% less energy than other leading ENERGY STAR® Certified freezers*. The FrostLess ULT offers advances that mitigate frost build-up while still delivering energy efficiency. Our FrostLess offers less frost build-up than our category-leading models, while maintaining the reliability and energy efficiency today’s laboratories require. Its new thermal insulation design helps prevent large amounts of frost build-up on the outside of the inner doors, reducing maintenance requirements.
*Based upon www.energystar.gov/productfinder/product/certified-lab-grade-refrigeration/ website accessed Jan 11, 2023
View all PHC Corporation products