MSDS Number: A5916 * * * * * Effective Date: 06/05/08 * * * * * Supercedes: 05/04/07 AMMONIUM HYDROXIDE (10 - 35% NH3)
MSDS   MATERIAL SAFETY DATA SHEET                  CHEMTREC: 800-424-9300 (USA)
              ---------------------------                    703-527-3887(Outside USA and Canada)
                                                                                              CANUTEC:    613-996-6666
From:    Mallinckrodt Baker, Inc
              222 Red School Lane
              Phillipsburg, NJ 08865                                  NOTE: Use CHEMTREC and CANUTEC
                                                                                                  phone numbers only in the event
Emergency Telephone Number: 908-859-2151                        of a chemical emergency.
All non-emergency questions should be directed to Customer Service (1-800-582-2537) for assistance.
M A L L I N C K R O D T                                      J. T. B A K E R
AMMONIUM HYDROXIDE (10 - 35% NH3)
1. Product Identification
Synonyms: Ammonium hydroxide solutions; ammonia aqueous; ammonia solutions
CAS No.: 1336-21-6
Molecular Weight: 35.05
Chemical Formula: NH4OH in H2O
Product Codes:
J.T. Baker: 4807, 5204, 5224, 5350, 5358, 5604, 5817, 5820, 5851, 5852, 5891, 5893, 5993, 7847, 9718, 9719, 9721, 9730, 9731, 9733, 9741, 9742, 9743
Mallinckrodt: 0124, 0127, 1177, 3256, 3258, 5318, 6665, H010, H893, H894, V592, V649, V893, XL002, XM-187, XM-189
2. Composition/Information on Ingredients
  Ingredient                                CAS No         Percent        Hazardous
  ---------------------------------------   ------------   ------------   ---------
  Ammonium Hydroxide                        1336-21-6        21 - 72%        Yes
  Water                                     7732-18-5        28 - 79%        No
  Contains between 10 and 35% ammonia.
3. Hazards Identification
Emergency Overview
--------------------------
POISON! DANGER! CORROSIVE. MAY BE FATAL IF SWALLOWED OR INHALED. MIST AND VAPOR CAUSE BURNS TO EVERY AREA OF CONTACT.
SAF-T-DATA(tm) Ratings (Provided here for your convenience)
-----------------------------------------------------------------------------------------------------------
Health Rating: 3 - Severe (Poison)
Flammability Rating: 0 - None
Reactivity Rating: 1 - Slight
Contact Rating: 4 - Extreme (Corrosive)
Lab Protective Equip: GOGGLES & SHIELD; LAB COAT & APRON; VENT HOOD; PROPER GLOVES
Storage Color Code: White Stripe (Store Separately)
-----------------------------------------------------------------------------------------------------------
Potential Health Effects
----------------------------------
Inhalation:
Vapors and mists cause irritation to the respiratory tract. Higher concentrations can cause burns, pulmonary edema and death. Brief exposure to 5000 ppm can be fatal.
Ingestion:
Toxic! May cause corrosion to the esophagus and stomach with perforation and peritonitis. Symptoms may include pain in the mouth, chest, and abdomen, with coughing, vomiting and collapse. Ingestion of as little as 3-4 mL may be fatal.
Skin Contact:
Causes irritation and burns to the skin.
Eye Contact:
Vapors cause irritation. Splashes cause severe pain, eye damage, and permanent blindness.
Chronic Exposure:
Repeated exposure may cause damage to the tissues of the mucous membranes, upper respiratory tract, eyes and skin.
Aggravation of Pre-existing Conditions:
Persons with pre-existing eye disorders or impaired respiratory function may be more susceptible to the effects of this material.
4. First Aid Measures
Inhalation:
Remove to fresh air. If not breathing, give artificial respiration. If breathing is difficult, give oxygen. Call a physician immediately.
Ingestion:
If swallowed, DO NOT INDUCE VOMITING. Give large quantities of water. Never give anything by mouth to an unconscious person. Get medical attention immediately.
Skin Contact:
Immediately flush skin with plenty of water for at least 15 minutes while removing contaminated clothing and shoes. Call a physician, immediately. Wash clothing before reuse.
Eye Contact:
Immediately flush eyes with gentle but large stream of water for at least 15 minutes, lifting lower and upper eyelids occasionally. Call a physician immediately. Immediate action is critical to minimize possibility of blindness.
5. Fire Fighting Measures
Fire:
Autoignition temperature: 651C (1204F)
Flammable limits in air % by volume:
lel: 16; uel: 25
Explosion:
Flammable vapors may accumulate in confined spaces.
Fire Extinguishing Media:
Use any means suitable for extinguishing surrounding fire. Use water spray to blanket fire, cool fire exposed containers, and to flush non-ignited spills or vapors away from fire.
Special Information:
In the event of a fire, wear full protective clothing and NIOSH-approved self-contained breathing apparatus with full facepiece operated in the pressure demand or other positive pressure mode.
6. Accidental Release Measures
Ventilate area of leak or spill. Keep unnecessary and unprotected people away from area of spill. Wear appropriate personal protective equipment as specified in Section 8. Contain and recover liquid when possible. Do not flush caustic residues to the sewer. Residues from spills can be diluted with water, neutralized with dilute acid such as acetic, hydrochloric or sulfuric. Absorb neutralized caustic residue on clay, vermiculite or other inert substance and package in a suitable container for disposal.
US Regulations (CERCLA) require reporting spills and releases to soil, water and air in excess of reportable quantities. The toll free number for the US Coast Guard National Response Center is (800) 424-8802.
J. T. Baker NEUTRACIT®-2 or BuCAIM® caustic neutralizers are recommended for spills of this product.
7. Handling and Storage
Keep in a tightly closed container, stored in a cool, dry, ventilated area. Protect against physical damage. Separate from incompatibilities. Store below 25C. Protect from direct sunlight. Containers of this material may be hazardous when empty since they retain product residues (vapors, liquid); observe all warnings and precautions listed for the product.
8. Exposure Controls/Personal Protection
Airborne Exposure Limits:
-OSHA Permissible Exposure Limit (PEL):
50 ppm (NH3)
-ACGIH Threshold Limit Value (TLV):
25 ppm (NH3) (TWA) 35 ppm (STEL)
Ventilation System:
A system of local and/or general exhaust is recommended to keep employee exposures below the Airborne Exposure Limits. Local exhaust ventilation is generally preferred because it can control the emissions of the contaminant at its source, preventing dispersion of it into the general work area. Please refer to the ACGIH document, Industrial Ventilation, A Manual of Recommended Practices, most recent edition, for details.
Personal Respirators (NIOSH Approved):
If the exposure limit is exceeded and engineering controls are not feasible, a full facepiece respirator with an ammonia/methylamine cartridge may be worn up to 50 times the exposure limit or the maximum use concentration specified by the appropriate regulatory agency or respirator supplier, whichever is lowest. For emergencies or instances where the exposure levels are not known, use a full-facepiece positive-pressure, air-supplied respirator. WARNING: Air purifying respirators do not protect workers in oxygen-deficient atmospheres.
Skin Protection:
Wear impervious protective clothing, including boots, gloves, lab coat, apron or coveralls, as appropriate, to prevent skin contact. Neoprene and nitrile rubber are recommended materials. Polyvinyl alcohol is not recommended.
Eye Protection:
Use chemical safety goggles and/or full face shield where dusting or splashing of solutions is possible. Maintain eye wash fountain and quick-drench facilities in work area.
9. Physical and Chemical Properties
Appearance:
Clear, colorless solution.
Odor:
Ammonia odor.
Solubility:
Infinitely soluble.
Specific Gravity:
0.9 (28% NH4OH)
pH:
13.8 (29% solution).
% Volatiles by volume @ 21C (70F):
No information found.
Boiling Point:
ca. 36C (ca. 97F)
Melting Point:
-72C (-98F)
Vapor Density (Air=1):
0.60 NH3
Vapor Pressure (mm Hg):
115 @ 20C for 10% solution; 580 @ 20C for 28% solution.
Evaporation Rate (BuAc=1):
No information found.
10. Stability and Reactivity
Stability:
Stable under ordinary conditions of use and storage.
Hazardous Decomposition Products:
Burning may produce ammonia, nitrogen oxides.
Hazardous Polymerization:
Will not occur.
Incompatibilities:
Acids, acrolein, dimethyl sulfate, halogens, silver nitrate, propylene oxide, nitromethane, silver oxide, silver permanganate, oleum, beta-propiolactone. Most common metals.
Conditions to Avoid:
Heat, sunlight, incompatibles, sources of ignition.
11. Toxicological Information
For ammonium hydroxide:
oral rat LD50: 350 mg/kg; eye, rabbit, standard Draize, 250 ug; severe, investigated as a mutagen.
For ammonia:
inhalation rat LC50: 2000 ppm/4-hr; investigated as a tumorigen, mutagen.
  --------\Cancer Lists\------------------------------------------------------
                                         ---NTP Carcinogen---
  Ingredient                             Known    Anticipated    IARC Category
  ------------------------------------   -----    -----------    -------------
  Ammonium Hydroxide (1336-21-6)          No          No            None
  Water (7732-18-5)                       No          No            None
12. Ecological Information
Environmental Fate:
This material is not expected to significantly bioaccumulate.
Environmental Toxicity:
24 Hr LC50 rainbow trout: 0.008 mg/L;
96 Hr LC50 fathead minnow: 8.2 mg/L;
48 Hr LC50 bluegill: 0.024 mg/L;
48 Hr EC50 water flea: 0.66 mg/L
13. Disposal Considerations
Whatever cannot be saved for recovery or recycling should be managed in an appropriate and approved waste facility. Although not a listed RCRA hazardous waste, this material may exhibit one or more characteristics of a hazardous waste and require appropriate analysis to determine specific disposal requirements. Processing, use or contamination of this product may change the waste management options. State and local disposal regulations may differ from federal disposal regulations. Dispose of container and unused contents in accordance with federal, state and local requirements.
14. Transport Information
Domestic (Land, D.O.T.)
-----------------------
Proper Shipping Name: AMMONIA SOLUTIONS (WITH 10-35% AMMONIA)
Hazard Class: 8
UN/NA: UN2672
Packing Group: III
Information reported for product/size: 385LB
International (Water, I.M.O.)
-----------------------------
Proper Shipping Name: AMMONIA SOLUTIONS (WITH 10-35% AMMONIA)
Hazard Class: 8
UN/NA: UN2672
Packing Group: III
Information reported for product/size: 385LB
15. Regulatory Information
WHMIS Classification: D1B, E
  --------\Chemical Inventory Status - Part 1\---------------------------------
  Ingredient                                       TSCA  EC   Japan  Australia
  -----------------------------------------------  ----  ---  -----  ---------
  Ammonium Hydroxide (1336-21-6)                    Yes  Yes   Yes      Yes
  Water (7732-18-5)                                 Yes  Yes   Yes      Yes
  --------\Chemical Inventory Status - Part 2\---------------------------------
                                                          --Canada--
  Ingredient                                       Korea  DSL   NDSL  Phil.
  -----------------------------------------------  -----  ---   ----  -----
  Ammonium Hydroxide (1336-21-6)                    Yes   Yes   No     Yes
  Water (7732-18-5)                                 Yes   Yes   No     Yes
  --------\Federal, State & International Regulations - Part 1\----------------
                                             -SARA 302-    ------SARA 313------
  Ingredient                                 RQ    TPQ     List  Chemical Catg.
  -----------------------------------------  ---   -----   ----  --------------
  Ammonium Hydroxide (1336-21-6)             No    No      No         No
  Water (7732-18-5)                          No    No      No         No
  --------\Federal, State & International Regulations - Part 2\----------------
                                                        -RCRA-    -TSCA-
  Ingredient                                 CERCLA     261.33     8(d)
  -----------------------------------------  ------     ------    ------
  Ammonium Hydroxide (1336-21-6)             1000       No         No
  Water (7732-18-5)                          No         No         No
Chemical Weapons Convention:  No     TSCA 12(b):  No     CDTA:  No
SARA 311/312:  Acute: Yes      Chronic: Yes  Fire: No  Pressure: No
Reactivity: No          (Mixture / Liquid)
Australian Hazchem Code: 2P
Poison Schedule: S6
WHMIS:
This MSDS has been prepared according to the hazard criteria of the Controlled Products Regulations (CPR) and the MSDS contains all of the information required by the CPR.
16. Other Information
NFPA Ratings: Health: 3 Flammability: 1 Reactivity: 0
Label Hazard Warning:
POISON! DANGER! CORROSIVE. MAY BE FATAL IF SWALLOWED OR INHALED. MIST AND VAPOR CAUSE BURNS TO EVERY AREA OF CONTACT.
Label Precautions:
Do not get in eyes, on skin, or on clothing.
Do not breathe vapor or mist.
Keep container closed.
Use only with adequate ventilation.
Wash thoroughly after handling.
Label First Aid:
If swallowed, DO NOT INDUCE VOMITING. Give large quantities of water. Never give anything by mouth to an unconscious person. If inhaled, remove to fresh air. If not breathing, give artificial respiration. If breathing is difficult, give oxygen. In case of contact, immediately flush eyes or skin with plenty of water for at least 15 minutes while removing contaminated clothing and shoes. Wash clothing before reuse. IMMEDIATE ACTION IS ESSENTIAL FOR EYE EXPOSURES. In all cases call a physician immediately.
Product Use:
Laboratory Reagent.
Revision Information:
MSDS Section(s) changed since last revision of document include: 15.
Disclaimer:
************************************************************************************************
Mallinckrodt Baker, Inc. provides the information contained herein in good faith but makes no representation as to its comprehensiveness or accuracy. This document is intended only as a guide to the appropriate precautionary handling of the material by a properly trained person using this product. Individuals receiving the information must exercise their independent judgment in determining its appropriateness for a particular purpose. MALLINCKRODT BAKER, INC. MAKES NO REPRESENTATIONS OR WARRANTIES, EITHER EXPRESS OR IMPLIED, INCLUDING WITHOUT LIMITATION ANY WARRANTIES OF MERCHANTABILITY, FITNESS FOR A PARTICULAR PURPOSE WITH RESPECT TO THE INFORMATION SET FORTH HEREIN OR THE PRODUCT TO WHICH THE INFORMATION REFERS. ACCORDINGLY, MALLINCKRODT BAKER, INC. WILL NOT BE RESPONSIBLE FOR DAMAGES RESULTING FROM USE OF OR RELIANCE UPON THIS INFORMATION.
************************************************************************************************
Prepared by: Environmental Health & Safety
Phone Number: (314) 654-1600 (U.S.A.)