MSDS Number: H4070 * * * * * Effective Date: 02/15/08 * * * * * Supercedes: 04/22/05 HYDROGEN PEROXIDE SOLUTION 3%
MSDS   MATERIAL SAFETY DATA SHEET                  CHEMTREC: 800-424-9300 (USA)
              ---------------------------                    703-527-3887(Outside USA and Canada)
                                                                                              CANUTEC:    613-996-6666
From:    Mallinckrodt Baker, Inc
              222 Red School Lane
              Phillipsburg, NJ 08865                                  NOTE: Use CHEMTREC and CANUTEC
                                                                                                  phone numbers only in the event
Emergency Telephone Number: 908-859-2151                        of a chemical emergency.
All non-emergency questions should be directed to Customer Service (1-800-582-2537) for assistance.
J. T. B A K E R
HYDROGEN PEROXIDE SOLUTION 3%
1. Product Identification
Synonyms: Hydrogen Dioxide Solution, 3%; Hydrogen Peroxide Topical Solution U.S.P
CAS No.: 7722-84-1
Molecular Weight: 34.01
Chemical Formula: H2O2 in aqueous solution (3%)
Product Codes: 2180, 2182
2. Composition/Information on Ingredients
  Ingredient                                CAS No         Percent        Hazardous
  ---------------------------------------   ------------   ------------   ---------
  Hydrogen Peroxide                         7722-84-1         2 - 4%         Yes
  Phenacetin                                62-44-2           < 0.05%        No
  Water                                     7732-18-5        96 - 98%        No
3. Hazards Identification
Emergency Overview
--------------------------
WARNING! MAY BE HARMFUL IF SWALLOWED. CAUSES EYE IRRITATION.
SAF-T-DATA(tm) Ratings (Provided here for your convenience)
-----------------------------------------------------------------------------------------------------------
Health Rating: 2 - Moderate
Flammability Rating: 0 - None
Reactivity Rating: 1 - Slight
Contact Rating: 2 - Moderate
Lab Protective Equip: GOGGLES; LAB COAT; VENT HOOD; PROPER GLOVES
Storage Color Code: Green (General Storage)
-----------------------------------------------------------------------------------------------------------
Potential Health Effects
----------------------------------
Inhalation:
Not expected to be a health hazard under normal conditions.
Ingestion:
Large oral doses may cause irritation and blistering to the mouth, throat, and abdomen. May also cause abdominal pain, vomiting, and diarrhea.
Skin Contact:
No adverse effects expected on intact skin. Contact on burn or open skin may cause stinging pain or irritation.
Eye Contact:
Causes irritation, redness, and pain.
Chronic Exposure:
No information found.
Aggravation of Pre-existing Conditions:
No information found.
4. First Aid Measures
Inhalation:
Not expected to require first aid measures.
Ingestion:
Give several glasses of water to drink to dilute. If large amounts were swallowed, get medical advice.
Skin Contact:
Not expected to require first aid measures. Wash exposed area with soap and water. Get medical advice if irritation develops.
Eye Contact:
Immediately flush eyes with plenty of water for at least 15 minutes, lifting lower and upper eyelids occasionally. Get medical attention immediately.
5. Fire Fighting Measures
Fire:
Not considered to be a fire hazard. Concentrated hydrogen peroxide (30%) is a strong oxidizer, but this dilute product does not present that hazard.
Explosion:
Not considered to be an explosion hazard. Drying of concentrated hydrogen peroxide on clothing or other combustible materials may cause fire or explosion.
Fire Extinguishing Media:
Use any means suitable for extinguishing surrounding fire.
Special Information:
In the event of a fire, wear full protective clothing and NIOSH-approved self-contained breathing apparatus with full facepiece operated in the pressure demand or other positive pressure mode.
6. Accidental Release Measures
Ventilate area of leak or spill. Wear appropriate personal protective equipment as specified in Section 8. Contain and recover liquid when possible. Collect liquid in an appropriate container or absorb with an inert material (e. g., vermiculite, dry sand, earth), and place in a chemical waste container. Do not use combustible materials, such as saw dust. Small amounts of residue may be flushed to sewer with plenty of water.
7. Handling and Storage
Store in a cool, well-ventilated dark area. Protect from freezing. Isolate from incompatible substances. Protect container from physical damage. Containers of this material may be hazardous when empty since they retain product residues (vapors, liquid); observe all warnings and precautions listed for the product.
8. Exposure Controls/Personal Protection
Airborne Exposure Limits:
-OSHA Permissible Exposure Limit (PEL):
1 ppm (TWA).
-ACGIH Threshold Limit Value (TLV):
1 ppm (TWA), A3: Animal carcinogen.
Ventilation System:
A system of local and/or general exhaust is recommended to keep employee exposures below the Airborne Exposure Limits. Local exhaust ventilation is generally preferred because it can control the emissions of the contaminant at its source, preventing dispersion of it into the general work area. Please refer to the ACGIH document, Industrial Ventilation, A Manual of Recommended Practices, most recent edition, for details.
Personal Respirators (NIOSH Approved):
Not expected to require personal respirator usage. If the exposure limit is exceeded, wear a supplied air, full-facepiece respirator, airlined hood, or full-facepiece self-contained breathing apparatus. This substance has unknown warning properties.
Skin Protection:
Wear protective gloves and clean body-covering clothing.
Eye Protection:
Use chemical safety goggles and/or a full face shield where splashing is possible. Maintain eye wash fountain and quick-drench facilities in work area.
9. Physical and Chemical Properties
Appearance:
Clear, colorless solution.
Odor:
Odorless.
Solubility:
Infinitely soluble.
Specific Gravity:
ca. 1.0
pH:
No information found.
% Volatiles by volume @ 21C (70F):
100
Boiling Point:
ca. 100C (ca. 212F)
Melting Point:
ca. 0C (ca. 32F)
Vapor Density (Air=1):
No information found.
Vapor Pressure (mm Hg):
No information found.
Evaporation Rate (BuAc=1):
No information found.
10. Stability and Reactivity
Stability:
Stable under ordinary conditions of use and storage.
Hazardous Decomposition Products:
Decomposes to water and oxygen.
Hazardous Polymerization:
Will not occur.
Incompatibilities:
Heat, reducing agents, organic materials, dirt, alkalis, rust, and many metals.
Conditions to Avoid:
Light, heat, incompatibles.
11. Toxicological Information
  --------\Cancer Lists\------------------------------------------------------
                                         ---NTP Carcinogen---
  Ingredient                             Known    Anticipated    IARC Category
  ------------------------------------   -----    -----------    -------------
  Hydrogen Peroxide (7722-84-1)           No          No              3
  Phenacetin (62-44-2)                    No          Yes            2A
  Water (7732-18-5)                       No          No            None
12. Ecological Information
Environmental Fate:
No information found.
Environmental Toxicity:
No information found.
13. Disposal Considerations
Dilute with water and flush to sewer if local ordinances allow, otherwise, whatever cannot be saved for recovery or recycling should be managed in an appropriate and approved waste disposal facility. Processing, use or contamination of this product may change the waste management options. State and local disposal regulations may differ from federal disposal regulations. Dispose of container and unused contents in accordance with federal, state and local requirements.
14. Transport Information
Not regulated.
15. Regulatory Information
  --------\Chemical Inventory Status - Part 1\---------------------------------
  Ingredient                                       TSCA  EC   Japan  Australia
  -----------------------------------------------  ----  ---  -----  ---------
  Hydrogen Peroxide (7722-84-1)                     Yes  Yes   Yes      Yes
  Phenacetin (62-44-2)                              Yes  Yes   Yes      Yes
  Water (7732-18-5)                                 Yes  Yes   Yes      Yes
  --------\Chemical Inventory Status - Part 2\---------------------------------
                                                          --Canada--
  Ingredient                                       Korea  DSL   NDSL  Phil.
  -----------------------------------------------  -----  ---   ----  -----
  Hydrogen Peroxide (7722-84-1)                     Yes   Yes   No     Yes
  Phenacetin (62-44-2)                              Yes   Yes   No     Yes
  Water (7732-18-5)                                 Yes   Yes   No     Yes
  --------\Federal, State & International Regulations - Part 1\----------------
                                             -SARA 302-    ------SARA 313------
  Ingredient                                 RQ    TPQ     List  Chemical Catg.
  -----------------------------------------  ---   -----   ----  --------------
  Hydrogen Peroxide (7722-84-1)              No    No      No         No
  Phenacetin (62-44-2)                       No    No      No         No
  Water (7732-18-5)                          No    No      No         No
  --------\Federal, State & International Regulations - Part 2\----------------
                                                        -RCRA-    -TSCA-
  Ingredient                                 CERCLA     261.33     8(d)
  -----------------------------------------  ------     ------    ------
  Hydrogen Peroxide (7722-84-1)              No         No         No
  Phenacetin (62-44-2)                       100        U187       No
  Water (7732-18-5)                          No         No         No
Chemical Weapons Convention:  No     TSCA 12(b):  No     CDTA:  No
SARA 311/312:  Acute: Yes      Chronic: No   Fire: No  Pressure: No
Reactivity: Yes         (Mixture / Liquid)
WARNING:
THIS PRODUCT CONTAINS A CHEMICAL(S) KNOWN TO THE STATE OF CALIFORNIA TO CAUSE CANCER.
Australian Hazchem Code: None allocated.
Poison Schedule: S5
WHMIS:
This MSDS has been prepared according to the hazard criteria of the Controlled Products Regulations (CPR) and the MSDS contains all of the information required by the CPR.
16. Other Information
NFPA Ratings: Health: 1 Flammability: 0 Reactivity: 1
Label Hazard Warning:
WARNING! MAY BE HARMFUL IF SWALLOWED. CAUSES EYE IRRITATION.
Label Precautions:
Avoid contact with eyes.
Keep container closed.
Wash thoroughly after handling.
Label First Aid:
In case of eye contact, immediately flush eyes with plenty of water for at least 15 minutes. Get medical attention. If swallowed, give large amounts of water to drink. Never give anything by mouth to an unconscious person. If large amounts were swallowed, get medical advice.
Product Use:
Laboratory Reagent.
Revision Information:
No Changes.
Disclaimer:
************************************************************************************************
Mallinckrodt Baker, Inc. provides the information contained herein in good faith but makes no representation as to its comprehensiveness or accuracy. This document is intended only as a guide to the appropriate precautionary handling of the material by a properly trained person using this product. Individuals receiving the information must exercise their independent judgment in determining its appropriateness for a particular purpose. MALLINCKRODT BAKER, INC. MAKES NO REPRESENTATIONS OR WARRANTIES, EITHER EXPRESS OR IMPLIED, INCLUDING WITHOUT LIMITATION ANY WARRANTIES OF MERCHANTABILITY, FITNESS FOR A PARTICULAR PURPOSE WITH RESPECT TO THE INFORMATION SET FORTH HEREIN OR THE PRODUCT TO WHICH THE INFORMATION REFERS. ACCORDINGLY, MALLINCKRODT BAKER, INC. WILL NOT BE RESPONSIBLE FOR DAMAGES RESULTING FROM USE OF OR RELIANCE UPON THIS INFORMATION.
************************************************************************************************
Prepared by: Environmental Health & Safety
Phone Number: (314) 654-1600 (U.S.A.)