MSDS Number: P5381 * * * * * Effective Date: 11/21/08 * * * * * Supercedes: 01/16/06 POTASSIUM ACETATE
MSDS   MATERIAL SAFETY DATA SHEET                  CHEMTREC: 800-424-9300 (USA)
              ---------------------------                    703-527-3887(Outside USA and Canada)
                                                                                              CANUTEC:    613-996-6666
From:    Mallinckrodt Baker, Inc
              222 Red School Lane
              Phillipsburg, NJ 08865                                  NOTE: Use CHEMTREC and CANUTEC
                                                                                                  phone numbers only in the event
Emergency Telephone Number: 908-859-2151                        of a chemical emergency.
All non-emergency questions should be directed to Customer Service (1-800-582-2537) for assistance.
M A L L I N C K R O D T                                      J. T. B A K E R
POTASSIUM ACETATE
1. Product Identification
Synonyms: Acetic acid, potassium salt
CAS No.: 127-08-2
Molecular Weight: 98.15
Chemical Formula: KC2H3O2
Product Codes:
J.T. Baker: 2912, 2914, 2915, 2917, 6072
Mallinckrodt: 6696, 6700, 7797
2. Composition/Information on Ingredients
  Ingredient                                CAS No         Percent        Hazardous
  ---------------------------------------   ------------   ------------   ---------
  Potassium Acetate                         127-08-2         99 - 100%       Yes
3. Hazards Identification
Emergency Overview
--------------------------
As part of good industrial and personal hygiene and safety procedure, avoid all unnecessary exposure to the chemical substance and ensure prompt removal from skin, eyes and clothing.
SAF-T-DATA(tm) Ratings (Provided here for your convenience)
-----------------------------------------------------------------------------------------------------------
Health Rating: 1 - Slight
Flammability Rating: 0 - None
Reactivity Rating: 0 - None
Contact Rating: 1 - Slight
Lab Protective Equip: GOGGLES; LAB COAT; PROPER GLOVES
Storage Color Code: Green (General Storage)
-----------------------------------------------------------------------------------------------------------
Potential Health Effects
----------------------------------
Inhalation:
May cause mild irritation to the respiratory tract.
Ingestion:
Extremely large oral dosages may produce gastrointestinal disturbances. Has diuretic effect.
Skin Contact:
No adverse effects expected. May cause mild irritation and redness.
Eye Contact:
No adverse effects expected. May cause mild irritation, possible reddening.
Chronic Exposure:
No adverse health effects expected.
Aggravation of Pre-existing Conditions:
No adverse health effects expected.
4. First Aid Measures
Inhalation:
Remove to fresh air. Get medical attention for any breathing difficulty.
Ingestion:
If large amounts were swallowed, give water to drink and get medical advice.
Skin Contact:
Wash exposed area with soap and water. Get medical advice if irritation develops.
Eye Contact:
Wash thoroughly with running water. Get medical advice if irritation develops.
5. Fire Fighting Measures
Fire:
Not considered to be a fire hazard.
Explosion:
Not considered to be an explosion hazard.
Fire Extinguishing Media:
Use any means suitable for extinguishing surrounding fire.
Special Information:
In the event of a fire, wear full protective clothing and NIOSH-approved self-contained breathing apparatus with full facepiece operated in the pressure demand or other positive pressure mode.
6. Accidental Release Measures
Ventilate area of leak or spill. Wear appropriate personal protective equipment as specified in Section 8. Spills: Sweep up and containerize for reclamation or disposal. Vacuuming or wet sweeping may be used to avoid dust dispersal.
7. Handling and Storage
Keep in a tightly closed container, stored in a cool, dry, ventilated area. Protect against physical damage. Containers of this material may be hazardous when empty since they retain product residues (dust, solids); observe all warnings and precautions listed for the product.
8. Exposure Controls/Personal Protection
Airborne Exposure Limits:
None established.
Ventilation System:
In general, dilution ventilation is a satisfactory health hazard control for this substance. However, if conditions of use create discomfort to the worker, a local exhaust system should be considered.
Personal Respirators (NIOSH Approved):
For conditions of use where exposure to dust or mist is apparent and engineering controls are not feasible, a particulate respirator (NIOSH type N95 or better filters) may be worn. If oil particles (e.g. lubricants, cutting fluids, glycerine, etc.) are present, use a NIOSH type R or P filter. For emergencies or instances where the exposure levels are not known, use a full-face positive-pressure, air-supplied respirator. WARNING: Air-purifying respirators do not protect workers in oxygen-deficient atmospheres.
Skin Protection:
Wear protective gloves and clean body-covering clothing.
Eye Protection:
Use chemical safety goggles. Maintain eye wash fountain and quick-drench facilities in work area.
9. Physical and Chemical Properties
Appearance:
White, deliquescent, crystalline powder.
Odor:
Odorless.
Solubility:
200 g/100 g water.
Specific Gravity:
1.57 @ 25C
pH:
9.7 Conc: 0.1M (solution)
% Volatiles by volume @ 21C (70F):
0
Boiling Point:
No information found.
Melting Point:
292C (558F)
Vapor Density (Air=1):
No information found.
Vapor Pressure (mm Hg):
No information found.
Evaporation Rate (BuAc=1):
No information found.
10. Stability and Reactivity
Stability:
Stable under ordinary conditions of use and storage.
Hazardous Decomposition Products:
May produce oxides of carbon and the contained metal.
Hazardous Polymerization:
Will not occur.
Incompatibilities:
Strong oxidizing agents.
Conditions to Avoid:
Moisture, heat, flames, ignition sources and incompatibles.
11. Toxicological Information
Oral rat LD50: 3250 mg/kg
  --------\Cancer Lists\------------------------------------------------------
                                         ---NTP Carcinogen---
  Ingredient                             Known    Anticipated    IARC Category
  ------------------------------------   -----    -----------    -------------
  Potassium Acetate (127-08-2)            No          No            None
12. Ecological Information
Environmental Fate:
No information found.
Environmental Toxicity:
No information found.
13. Disposal Considerations
Whatever cannot be saved for recovery or recycling should be managed in an appropriate and approved waste disposal facility. Processing, use or contamination of this product may change the waste management options. State and local disposal regulations may differ from federal disposal regulations. Dispose of container and unused contents in accordance with federal, state and local requirements.
14. Transport Information
Not regulated.
15. Regulatory Information
  --------\Chemical Inventory Status - Part 1\---------------------------------
  Ingredient                                       TSCA  EC   Japan  Australia
  -----------------------------------------------  ----  ---  -----  ---------
  Potassium Acetate (127-08-2)                      Yes  Yes   Yes      Yes
  --------\Chemical Inventory Status - Part 2\---------------------------------
                                                          --Canada--
  Ingredient                                       Korea  DSL   NDSL  Phil.
  -----------------------------------------------  -----  ---   ----  -----
  Potassium Acetate (127-08-2)                      Yes   Yes   No     Yes
  --------\Federal, State & International Regulations - Part 1\----------------
                                             -SARA 302-    ------SARA 313------
  Ingredient                                 RQ    TPQ     List  Chemical Catg.
  -----------------------------------------  ---   -----   ----  --------------
  Potassium Acetate (127-08-2)               No    No      No         No
  --------\Federal, State & International Regulations - Part 2\----------------
                                                        -RCRA-    -TSCA-
  Ingredient                                 CERCLA     261.33     8(d)
  -----------------------------------------  ------     ------    ------
  Potassium Acetate (127-08-2)               No         No         No
Chemical Weapons Convention:  No     TSCA 12(b):  No     CDTA:  No
SARA 311/312:  Acute: No       Chronic: No   Fire: No  Pressure: No
Reactivity: No          (Pure / Solid)
Australian Hazchem Code: None allocated.
Poison Schedule: None allocated.
WHMIS:
This MSDS has been prepared according to the hazard criteria of the Controlled Products Regulations (CPR) and the MSDS contains all of the information required by the CPR.
16. Other Information
NFPA Ratings: Health: 1 Flammability: 0 Reactivity: 0
Label Hazard Warning:
As part of good industrial and personal hygiene and safety procedure, avoid all unnecessary exposure to the chemical substance and ensure prompt removal from skin, eyes and clothing.
Label Precautions:
None.
Label First Aid:
Not applicable.
Product Use:
Laboratory Reagent.
Revision Information:
No Changes.
Disclaimer:
************************************************************************************************
Mallinckrodt Baker, Inc. provides the information contained herein in good faith but makes no representation as to its comprehensiveness or accuracy. This document is intended only as a guide to the appropriate precautionary handling of the material by a properly trained person using this product. Individuals receiving the information must exercise their independent judgment in determining its appropriateness for a particular purpose. MALLINCKRODT BAKER, INC. MAKES NO REPRESENTATIONS OR WARRANTIES, EITHER EXPRESS OR IMPLIED, INCLUDING WITHOUT LIMITATION ANY WARRANTIES OF MERCHANTABILITY, FITNESS FOR A PARTICULAR PURPOSE WITH RESPECT TO THE INFORMATION SET FORTH HEREIN OR THE PRODUCT TO WHICH THE INFORMATION REFERS. ACCORDINGLY, MALLINCKRODT BAKER, INC. WILL NOT BE RESPONSIBLE FOR DAMAGES RESULTING FROM USE OF OR RELIANCE UPON THIS INFORMATION.
************************************************************************************************
Prepared by: Environmental Health & Safety
Phone Number: (314) 654-1600 (U.S.A.)