MSDS Number: S2670 * * * * * Effective Date: 02/16/06 * * * * * Supercedes: 02/16/06 SODIUM ACETATE ANHYDROUS
MSDS   MATERIAL SAFETY DATA SHEET                  CHEMTREC: 800-424-9300 (USA)
              ---------------------------                    703-527-3887(Outside USA and Canada)
                                                                                              CANUTEC:    613-996-6666
From:    Mallinckrodt Baker, Inc
              222 Red School Lane
              Phillipsburg, NJ 08865                                  NOTE: Use CHEMTREC and CANUTEC
                                                                                                  phone numbers only in the event
Emergency Telephone Number: 908-859-2151                        of a chemical emergency.
All non-emergency questions should be directed to Customer Service (1-800-582-2537) for assistance.
M A L L I N C K R O D T                                      J. T. B A K E R
SODIUM ACETATE ANHYDROUS
1. Product Identification
Synonyms: Sodium acetate anhydrous; Acetic acid, sodium salt
CAS No.: 127-09-3
Molecular Weight: 82.03
Chemical Formula: CH3COONa
Product Codes:
J.T. Baker: 3470, 3472, 3473, 3474
Mallinckrodt: 7372, 7800
2. Composition/Information on Ingredients
  Ingredient                                CAS No         Percent        Hazardous
  ---------------------------------------   ------------   ------------   ---------
  Sodium Acetate                            127-09-3         90 - 100%       Yes
3. Hazards Identification
Emergency Overview
--------------------------
CAUTION! MAY CAUSE IRRITATION TO SKIN, EYES, AND RESPIRATORY TRACT.
SAF-T-DATA(tm) Ratings (Provided here for your convenience)
-----------------------------------------------------------------------------------------------------------
Health Rating: 1 - Slight
Flammability Rating: 1 - Slight
Reactivity Rating: 1 - Slight
Contact Rating: 2 - Moderate
Lab Protective Equip: GOGGLES; LAB COAT; VENT HOOD; PROPER GLOVES
Storage Color Code: Green (General Storage)
-----------------------------------------------------------------------------------------------------------
Potential Health Effects
----------------------------------
Inhalation:
May cause irritation to the respiratory tract. Symptoms may include coughing, sore throat, labored breathing, and chest pain.
Ingestion:
Large doses may produce abdominal pain, nausea, and vomiting.
Skin Contact:
May cause irritation with redness and pain.
Eye Contact:
May cause irritation, redness and pain.
Chronic Exposure:
No information found.
Aggravation of Pre-existing Conditions:
No information found.
4. First Aid Measures
Inhalation:
Remove to fresh air. Get medical attention for any breathing difficulty.
Ingestion:
Give several glasses of water to drink to dilute. If large amounts were swallowed, get medical advice.
Skin Contact:
Immediately flush skin with plenty of water for at least 15 minutes. Remove contaminated clothing and shoes. Wash clothing before reuse. Thoroughly clean shoes before reuse. Get medical attention if irritation develops.
Eye Contact:
Immediately flush eyes with plenty of water for at least 15 minutes, lifting upper and lower eyelids occasionally. Get medical attention if irritation persists.
5. Fire Fighting Measures
Fire:
Autoignition temperature: 611C (1132F)
Combustible. May pose a fire hazard when exposed to elevated temperatures or by contact with an ignition source.
Explosion:
Fine dust dispersed in air in sufficient concentrations, and in the presence of an ignition source is a potential dust explosion hazard.
Fire Extinguishing Media:
Water spray, dry chemical, alcohol foam, or carbon dioxide.
Special Information:
In the event of a fire, wear full protective clothing and NIOSH-approved self-contained breathing apparatus with full facepiece operated in the pressure demand or other positive pressure mode.
6. Accidental Release Measures
Remove all sources of ignition. Ventilate area of leak or spill. Wear appropriate personal protective equipment as specified in Section 8. Spills: Clean up spills in a manner that does not disperse dust into the air. Use non-sparking tools and equipment. Reduce airborne dust and prevent scattering by moistening with water. Pick up spill for recovery or disposal and place in a closed container. Small amounts of residue may be flushed to sewer with plenty of water.
7. Handling and Storage
Keep in a tightly closed container, stored in a cool, dry, ventilated area. Protect against physical damage. Isolate from any source of heat or ignition. Containers of this material may be hazardous when empty since they retain product residues (dust, solids); observe all warnings and precautions listed for the product.
8. Exposure Controls/Personal Protection
Airborne Exposure Limits:
None established.
Ventilation System:
A system of local and/or general exhaust is recommended to keep employee exposures as low as possible. Local exhaust ventilation is generally preferred because it can control the emissions of the contaminant at its source, preventing dispersion of it into the general work area. Please refer to the ACGIH document, Industrial Ventilation, A Manual of Recommended Practices, most recent edition, for details.
Personal Respirators (NIOSH Approved):
For conditions of use where exposure to dust or mist is apparent and engineering controls are not feasible, a particulate respirator (NIOSH type N95 or better filters) may be worn. If oil particles (e.g. lubricants, cutting fluids, glycerine, etc.) are present, use a NIOSH type R or P filter. For emergencies or instances where the exposure levels are not known, use a full-face positive-pressure, air-supplied respirator. WARNING: Air-purifying respirators do not protect workers in oxygen-deficient atmospheres.
Skin Protection:
Wear protective gloves and clean body-covering clothing.
Eye Protection:
Use chemical safety goggles. Maintain eye wash fountain and quick-drench facilities in work area.
9. Physical and Chemical Properties
Appearance:
White hygroscopic crystals.
Odor:
Odorless.
Solubility:
119 g/100 g cold water
Density:
1.53
pH:
No information found.
% Volatiles by volume @ 21C (70F):
0
Boiling Point:
Not applicable.
Melting Point:
324C (615F)
Vapor Density (Air=1):
No information found.
Vapor Pressure (mm Hg):
No information found.
Evaporation Rate (BuAc=1):
No information found.
10. Stability and Reactivity
Stability:
Stable under ordinary conditions of use and storage. Hygroscopic powder.
Hazardous Decomposition Products:
Emits fumes of acetic acid upon heating and on contact with strong acids.
Hazardous Polymerization:
Will not occur.
Incompatibilities:
Nitric acid, fluoride, potassium nitrate, strong oxidizers and diketene.
Conditions to Avoid:
Heat, flames, ignition sources and incompatibles.
11. Toxicological Information
Oral rat LD50: 3530 mg/kg; inhalation rat LC50: > 30 gm/m3; skin rabbit LD50: > 10 mg/kg; Irritation Data, standard Draize: Skin rabbit 500 mg/24H, mild; standard Draize, Eye rabbit 10 mg, mild. Investigated as a mutagen.
  --------\Cancer Lists\------------------------------------------------------
                                         ---NTP Carcinogen---
  Ingredient                             Known    Anticipated    IARC Category
  ------------------------------------   -----    -----------    -------------
  Sodium Acetate (127-09-3)               No          No            None
12. Ecological Information
Environmental Fate:
No information found.
Environmental Toxicity:
Freshwater Fish Species Data: 24 Hr LC50 Lepomis macrochirus: 5000 mg/L
Microtox Data: 18 Hr EC50 Pseudomonas putida: 7200 mg/L
Water Flea Data: 48 Hr EC50 water flea: 5800 mg/L
13. Disposal Considerations
Whatever cannot be saved for recovery or recycling should be managed in an appropriate and approved waste disposal facility. Processing, use or contamination of this product may change the waste management options. State and local disposal regulations may differ from federal disposal regulations. Dispose of container and unused contents in accordance with federal, state and local requirements.
14. Transport Information
Not regulated.
15. Regulatory Information
  --------\Chemical Inventory Status - Part 1\---------------------------------
  Ingredient                                       TSCA  EC   Japan  Australia
  -----------------------------------------------  ----  ---  -----  ---------
  Sodium Acetate (127-09-3)                         Yes  Yes   Yes      Yes
  --------\Chemical Inventory Status - Part 2\---------------------------------
                                                          --Canada--
  Ingredient                                       Korea  DSL   NDSL  Phil.
  -----------------------------------------------  -----  ---   ----  -----
  Sodium Acetate (127-09-3)                         Yes   Yes   No     Yes
  --------\Federal, State & International Regulations - Part 1\----------------
                                             -SARA 302-    ------SARA 313------
  Ingredient                                 RQ    TPQ     List  Chemical Catg.
  -----------------------------------------  ---   -----   ----  --------------
  Sodium Acetate (127-09-3)                  No    No      No         No
  --------\Federal, State & International Regulations - Part 2\----------------
                                                        -RCRA-    -TSCA-
  Ingredient                                 CERCLA     261.33     8(d)
  -----------------------------------------  ------     ------    ------
  Sodium Acetate (127-09-3)                  No         No         No
Chemical Weapons Convention:  No     TSCA 12(b):  No     CDTA:  No
SARA 311/312:  Acute: Yes      Chronic: No   Fire: No  Pressure: No
Reactivity: No          (Pure / Solid)
Australian Hazchem Code: None allocated.
Poison Schedule: None allocated.
WHMIS:
This MSDS has been prepared according to the hazard criteria of the Controlled Products Regulations (CPR) and the MSDS contains all of the information required by the CPR.
16. Other Information
NFPA Ratings: Health: 1 Flammability: 1 Reactivity: 0
Label Hazard Warning:
CAUTION! MAY CAUSE IRRITATION TO SKIN, EYES, AND RESPIRATORY TRACT.
Label Precautions:
Avoid contact with eyes, skin and clothing.
Avoid breathing dust.
Wash thoroughly after handling.
Keep container closed.
Use with adequate ventilation.
Label First Aid:
If inhaled, remove to fresh air. Get medical attention for any breathing difficulty. In case of contact, immediately flush eyes or skin with plenty of water for at least 15 minutes. Get medical attention if irritation develops or persists.
Product Use:
Laboratory Reagent.
Revision Information:
MSDS Section(s) changed since last revision of document include: 3, 12.
Disclaimer:
************************************************************************************************
Mallinckrodt Baker, Inc. provides the information contained herein in good faith but makes no representation as to its comprehensiveness or accuracy. This document is intended only as a guide to the appropriate precautionary handling of the material by a properly trained person using this product. Individuals receiving the information must exercise their independent judgment in determining its appropriateness for a particular purpose. MALLINCKRODT BAKER, INC. MAKES NO REPRESENTATIONS OR WARRANTIES, EITHER EXPRESS OR IMPLIED, INCLUDING WITHOUT LIMITATION ANY WARRANTIES OF MERCHANTABILITY, FITNESS FOR A PARTICULAR PURPOSE WITH RESPECT TO THE INFORMATION SET FORTH HEREIN OR THE PRODUCT TO WHICH THE INFORMATION REFERS. ACCORDINGLY, MALLINCKRODT BAKER, INC. WILL NOT BE RESPONSIBLE FOR DAMAGES RESULTING FROM USE OF OR RELIANCE UPON THIS INFORMATION.
************************************************************************************************
Prepared by: Environmental Health & Safety
Phone Number: (314) 654-1600 (U.S.A.)