MSDS Number: Z7071 * * * * * Effective Date: 06/11/07 * * * * * Supercedes: 05/07/07 Speedisk® Column H2O-Philic DVB
MSDS   MATERIAL SAFETY DATA SHEET                  CHEMTREC: 800-424-9300 (USA)
              ---------------------------                    703-527-3887(Outside USA and Canada)
                                                                                              CANUTEC:    613-996-6666
From:    Mallinckrodt Baker, Inc
              222 Red School Lane
              Phillipsburg, NJ 08865                                  NOTE: Use CHEMTREC and CANUTEC
                                                                                                  phone numbers only in the event
Emergency Telephone Number: 908-859-2151                        of a chemical emergency.
All non-emergency questions should be directed to Customer Service (1-800-582-2537) for assistance.
J. T. B A K E R
Speedisk® Column H2O-Philic DVB
1. Product Identification
Synonyms: None
CAS No.: Not available.
Molecular Weight: Not available.
Chemical Formula: Not available.
Product Codes: 8077, 8082, 8108
2. Composition/Information on Ingredients
  Ingredient                                CAS No         Percent        Hazardous
  ---------------------------------------   ------------   ------------   ---------
  Hydrophilized DVB                         N/A                100%          No
3. Hazards Identification
Emergency Overview
--------------------------
As part of good industrial and personal hygiene and safety procedure, avoid all unnecessary exposure to the chemical substance and ensure prompt removal from skin, eyes and clothing.
SAF-T-DATA(tm) Ratings (Provided here for your convenience)
-----------------------------------------------------------------------------------------------------------
Health Rating: 1 - Slight
Flammability Rating: 0 - None
Reactivity Rating: 1 - Slight
Contact Rating: 1 - Slight
Lab Protective Equip: GOGGLES; LAB COAT; PROPER GLOVES
Storage Color Code: Green (General Storage)
-----------------------------------------------------------------------------------------------------------
Potential Health Effects
----------------------------------
NOTE: The hazard information listed is for the chromatography packing contained within the pre-packed chromatography column. Information on the human health effects from exposure to this substance is limited.
Inhalation:
Not expected to be a health hazard. Vapors are unlikely due to physical properties.
Ingestion:
No adverse effects expected. Extremely large oral dosages may produce gastrointestinal disturbances.
Skin Contact:
Not expected to be a health hazard from skin exposure. Skin absorption is unlikely due to physical properties.
Eye Contact:
Solid or dust may cause irritation or corneal injury due to mechanical action.
Chronic Exposure:
No adverse health effects expected.
Aggravation of Pre-existing Conditions:
No information found.
4. First Aid Measures
Inhalation:
Not expected to require first aid measures. Remove to fresh air. Get medical attention for any breathing difficulty.
Ingestion:
Not expected to require first aid measures. If large amounts were swallowed, give water to drink and get medical advice.
Skin Contact:
Not expected to require first aid measures. Wash exposed area with soap and water. Get medical advice if irritation develops.
Eye Contact:
Not expected to require first aid measures. Wash thoroughly with running water. Get medical advice if irritation develops.
5. Fire Fighting Measures
Fire:
Not considered to be a fire hazard.
Explosion:
Not considered to be an explosion hazard.
Fire Extinguishing Media:
Dry chemical.
Special Information:
In the event of a fire, wear full protective clothing and NIOSH-approved self-contained breathing apparatus with full facepiece operated in the pressure demand or other positive pressure mode.
6. Accidental Release Measures
Remove all sources of ignition. Caution! Floor and other surfaces may be slippery. Ventilate area of leak or spill. Wear appropriate personal protective equipment as specified in Section 8. Spills: Sweep up and containerize for reclamation or disposal. Vacuuming or wet sweeping may be used to avoid dust dispersal.
7. Handling and Storage
Keep in a tightly closed container, stored in a cool, dry, ventilated area. Protect against physical damage. Properly designed equipment is vital if these ion exchange resins are to be used in conjunction with strong oxidizing agents such as nitric acid to prevent a rapid build-up of pressure and possible explosion. Consult a source knowledgeable in the handling of these materials before proceeding. Do not pack column with dry ion exchange resins. Dry beads expand when wetted; this expansion can cause glass columns to shatter. Containers of this material may be hazardous when empty since they retain product residues (dust, solids); observe all warnings and precautions listed for the product.
8. Exposure Controls/Personal Protection
Airborne Exposure Limits:
None established.
Ventilation System:
Not expected to require any special ventilation.
Personal Respirators (NIOSH Approved):
Not expected to require personal respirator usage.
Skin Protection:
Wear protective gloves and clean body-covering clothing.
Eye Protection:
Use chemical safety goggles. Maintain eye wash fountain and quick-drench facilities in work area.
9. Physical and Chemical Properties
Appearance:
White to slightly yellow or tan particle bed or suspension prepacked inside a plastic column.
Odor:
Odorless.
Solubility:
Insoluble
Density:
No information found.
pH:
No information found.
% Volatiles by volume @ 21C (70F):
No information found.
Boiling Point:
Not applicable.
Melting Point:
No information found.
Vapor Density (Air=1):
Not applicable.
Vapor Pressure (mm Hg):
Not applicable.
Evaporation Rate (BuAc=1):
No information found.
10. Stability and Reactivity
Stability:
Stable under ordinary conditions of use and storage.
Hazardous Decomposition Products:
Oxides of carbon and possibly divinylbenzene monomers.
Hazardous Polymerization:
Will not occur.
Incompatibilities:
Avoid contact with strong oxidizing agents, particularly concentrated nitric acid.
Oxidizing agents such as nitric acid attack organic ion exchange resin under certain conditions. This could lead to anything from a slightly degraded resin up to an explosive reaction. Before using strong oxidizing agents, consult sources knowledgeable in handling such materials.
Conditions to Avoid:
Heat, flames, ignition sources and incompatibles.
11. Toxicological Information
No LD50/LC50 information found relating to normal routes of occupational exposure.
  --------\Cancer Lists\------------------------------------------------------
                                         ---NTP Carcinogen---
  Ingredient                             Known    Anticipated    IARC Category
  ------------------------------------   -----    -----------    -------------
  Hydrophilized DVB                       No          No            None
12. Ecological Information
Environmental Fate:
No information found.
Environmental Toxicity:
No information found.
13. Disposal Considerations
Whatever cannot be saved for recovery or recycling should be managed in an appropriate and approved waste disposal facility. Processing, use or contamination of this product may change the waste management options. State and local disposal regulations may differ from federal disposal regulations. Dispose of container and unused contents in accordance with federal, state and local requirements.
14. Transport Information
Not regulated.
15. Regulatory Information
  --------\Chemical Inventory Status - Part 1\---------------------------------
  Ingredient                                       TSCA  EC   Japan  Australia
  -----------------------------------------------  ----  ---  -----  ---------
  Hydrophilized DVB                               Exempt No    No       No
  --------\Chemical Inventory Status - Part 2\---------------------------------
                                                          --Canada--
  Ingredient                                       Korea  DSL   NDSL  Phil.
  -----------------------------------------------  -----  ---   ----  -----
  Hydrophilized DVB                                 No    No    No     No
  --------\Federal, State & International Regulations - Part 1\----------------
                                             -SARA 302-    ------SARA 313------
  Ingredient                                 RQ    TPQ     List  Chemical Catg.
  -----------------------------------------  ---   -----   ----  --------------
  Hydrophilized DVB                          No    No      No         No
  --------\Federal, State & International Regulations - Part 2\----------------
                                                        -RCRA-    -TSCA-
  Ingredient                                 CERCLA     261.33     8(d)
  -----------------------------------------  ------     ------    ------
  Hydrophilized DVB                          No         No         No
Chemical Weapons Convention:  No     TSCA 12(b):  No     CDTA:  No
SARA 311/312:  Acute: No       Chronic: No   Fire: No  Pressure: No
Reactivity: No          (Pure / Solid)
Australian Hazchem Code: None allocated.
Poison Schedule: None allocated.
WHMIS:
This MSDS has been prepared according to the hazard criteria of the Controlled Products Regulations (CPR) and the MSDS contains all of the information required by the CPR.
16. Other Information
NFPA Ratings: Health: 0 Flammability: 0 Reactivity: 0
Label Hazard Warning:
As part of good industrial and personal hygiene and safety procedure, avoid all unnecessary exposure to the chemical substance and ensure prompt removal from skin, eyes and clothing.
Label Precautions:
INTENDED FOR R & D USE ONLY.
NOT ON THE TSCA INVENTORY.
Label First Aid:
Not applicable.
Product Use:
Research and Development.
Revision Information:
MSDS Section(s) changed since last revision of document include: 3.
Disclaimer:
************************************************************************************************
Mallinckrodt Baker, Inc. provides the information contained herein in good faith but makes no representation as to its comprehensiveness or accuracy. This document is intended only as a guide to the appropriate precautionary handling of the material by a properly trained person using this product. Individuals receiving the information must exercise their independent judgment in determining its appropriateness for a particular purpose. MALLINCKRODT BAKER, INC. MAKES NO REPRESENTATIONS OR WARRANTIES, EITHER EXPRESS OR IMPLIED, INCLUDING WITHOUT LIMITATION ANY WARRANTIES OF MERCHANTABILITY, FITNESS FOR A PARTICULAR PURPOSE WITH RESPECT TO THE INFORMATION SET FORTH HEREIN OR THE PRODUCT TO WHICH THE INFORMATION REFERS. ACCORDINGLY, MALLINCKRODT BAKER, INC. WILL NOT BE RESPONSIBLE FOR DAMAGES RESULTING FROM USE OF OR RELIANCE UPON THIS INFORMATION.
************************************************************************************************
Prepared by: Environmental Health & Safety
Phone Number: (314) 654-1600 (U.S.A.)